Benzoin methyl ether 98%

Supplier: Ambeed
Danger

Synonyms: 1,2-Diphenyl-2-methoxyethan-1-one, 2-Methoxy-1,2-diphenylethanone

A135551-1G A135551-25G A135551-5G
77258-632EA 8.24 USD
77258-632 77258-634 77258-636
Benzoin methyl ether 98%
Benzoin methyl ether

Precaución: for Research Use Only

Formula: C₁₅H₁₄O₂
MW: 226.27 g/mol
MDL Number: MFCD00008492
CAS Number: 3524-62-7
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR