4'-Bromo-3'-nitroacetophenone 98%

Supplier: Ambeed
Warning

Synonyms: 1-(4-Bromo-3-nitrophenyl)ethan-1-one, 1-(4-Bromo-3-nitrophenyl)ethanone

A135550-100G A135550-25G A135550-5G A135550-1G A135550-250MG
77272-212EA 242 USD
77272-212 77272-214 77272-216 77615-082 77747-967
4'-Bromo-3'-nitroacetophenone 98%
4'-Bromo-3'-nitroacetophenone

Precaución: for Research Use Only

Formula: C₈H₆BrNO₃
MDL Number: MFCD00016985
CAS Number: 18640-58-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR