2,2':6',2''-Terpyridine-4'-carboxylic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
[2,2':6',2''-Terpyridine]-4'-carboxylic acid
A135533-1G
A135533-250MG
A135533-25G
A135533-5G
77246-710EA
35.02
USD
77246-710
77246-712
77246-714
77246-716
2,2':6',2''-Terpyridine-4'-carboxylic acid 97%
2,2':6',2''-Terpyridine-4'-carboxylic acid
Precaución: for Research Use Only
Formula:
C₁₆H₁₁N₃O₂ MW: 277.283 g/mol |
MDL Number:
MFCD04114290 CAS Number: 148332-36-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...