WZB117 99+%

Supplier: Ambeed
Warning

Synonyms: (3-fluoro-1,2-phenylene)ester, 3-hydroxy-benzoic acid, 3-Fluoro-1,2-phenylene bis(3-hydroxybenzoate)

A135447-100MG A135447-10MG A135447-1MG A135447-250MG A135447-25MG A135447-50MG A135447-5MG
77258-618EA 95 USD
77258-618 77258-620 77258-622 77258-624 77258-626 77258-628 77258-630
WZB117 99+%
WZB117

Precaución: for Research Use Only

Formula: C₂₀H₁₃FO₆
MDL Number: MFCD24387113
CAS Number: 1223397-11-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR