6-Fluoro-1-indanone 97%

Supplier: Ambeed
Warning

A135141-100G A135141-1G A135141-250MG A135141-25G A135141-5G
77258-602EA 193.64 USD
77258-602 77258-604 77258-606 77258-608 77258-610
6-Fluoro-1-indanone 97%
6-Fluoro-1-indanone

Precaución: for Research Use Only

Formula: C₉H₇FO
MW: 150.15 g/mol
MDL Number: MFCD01318147
CAS Number: 1481-32-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR