Ethyl 3-fluoro-4-(trifluoromethyl)benzoate 95%

Supplier: Ambeed
Warning

A135091-1G A135091-250MG
77258-598EA 48 USD
77258-598 77258-600
Ethyl 3-fluoro-4-(trifluoromethyl)benzoate 95%
Ethyl 3-fluoro-4-(trifluoromethyl)benzoate

Precaución: for Research Use Only

Formula: C₁₀H₈F₄O₂
CAS Number: 773135-31-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR