5-Bromo-7-ethyl-1H-indazole 95%

Supplier: Ambeed
Warning

A135010-100MG A135010-1G A135010-250MG
77234-684EA 42.23 USD
77234-684 77234-686 77234-688
5-Bromo-7-ethyl-1H-indazole 95%
5-Bromo-7-ethyl-1H-indazole

Precaución: for Research Use Only

Formula: C₉H₉BrN₂
MW: 225.085 g/mol
MDL Number: MFCD11044594
CAS Number: 635712-49-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR