3-(2-Iodophenyl)propanoic acid 97%

Supplier: Ambeed
Warning

A134851-1G A134851-250MG A134851-5G
77272-162EA 46.35 USD
77272-162 77272-164 77272-166
3-(2-Iodophenyl)propanoic acid 97%
3-(2-Iodophenyl)propanoic acid

Precaución: for Research Use Only

Formula: C₉H₉IO₂
MW: 276.071 g/mol
MDL Number: MFCD00040789
CAS Number: 96606-95-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR