8-Chloro-7-methoxyquinoline 97%

Supplier: Ambeed
Warning

A134803-100MG A134803-1G A134803-250MG
77234-668EA 142.14 USD
77234-668 77234-670 77234-672
8-Chloro-7-methoxyquinoline 97%
8-Chloro-7-methoxyquinoline

Precaución: for Research Use Only

Formula: C₁₀H₈ClNO
MW: 193.63 g/mol
MDL Number: MFCD28337090
CAS Number: 1422978-91-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR