Ethyl 4-amino-6-chloro-5-iodonicotinate 97%
Supplier: Ambeed
|
Warning
|
A134727-100MG
A134727-250MG
A134727-5G
77258-590EA
38.11
USD
77258-590
77258-592
77747-964
Ethyl 4-amino-6-chloro-5-iodonicotinate 97%
Ethyl 4-amino-6-chloro-5-iodonicotinate
Precaución: for Research Use Only
Formula:
C₈H₈ClIN₂O₂ MW: 326.51 g/mol |
MDL Number:
MFCD28559209 CAS Number: 1935952-84-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...