3-(Aminomethyl)benzoic acid hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: 3-(Aminomethyl)benzoic acid, HCl

A134699-100MG A134699-10G A134699-250MG A134699-25G A134699-5G
77246-678EA 6.18 USD
77246-678 77246-680 77246-682 77246-684 77246-686
3-(Aminomethyl)benzoic acid hydrochloride 97%
3-(Aminomethyl)benzoic acid hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₀ClNO₂
MW: 187.62 g/mol
MDL Number: MFCD03791117
CAS Number: 876-03-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR