5-Bromo-4-chloro-6-iodothieno[2,3-d]pyrimidine 97%

Supplier: Ambeed
Warning

A134652-1G A134652-250MG A134652-5G
77258-584EA 43 USD
77258-584 77258-586 77258-588
5-Bromo-4-chloro-6-iodothieno[2,3-d]pyrimidine 97%
5-Bromo-4-chloro-6-iodothieno[2,3-d]pyrimidine

Precaución: for Research Use Only

Formula: C₆HBrClIN₂S
MDL Number: MFCD30728471
CAS Number: 1799610-89-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR