ω-Bromo-3,4-dichloroacetophenone 97%

Supplier: Ambeed
Danger

A134603-100G A134603-10G A134603-1G A134603-5G
77272-154EA 226.01 USD
77272-154 77272-156 77272-158 77272-160
ω-Bromo-3,4-dichloroacetophenone 97%
ω-Bromo-3,4-dichloroacetophenone

Precaución: for Research Use Only

Formula: C₈H₅BrCl₂O
MDL Number: MFCD00051581
CAS Number: 2632-10-2
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR