1-Methyl-5-(4-(trifluoromethoxy)phenyl)-1H-pyrrole-2-carboxylic acid 95%

Supplier: Ambeed
Warning

A134575-1G A134575-250MG
77258-580EA 200.85 USD
77258-580 77258-582
1-Methyl-5-(4-(trifluoromethoxy)phenyl)-1H-pyrrole-2-carboxylic acid 95%
1-Methyl-5-(4-(trifluoromethoxy)phenyl)-1H-pyrrole-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₀F₃NO₃
MW: 285.22 g/mol
MDL Number: MFCD28252201
CAS Number: 1864060-29-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR