4,5-Difluoro-2-iodobenzoic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4,5-Difluoro-2-Iodobenzoic acid

A134416-100G A134416-10G A134416-1G A134416-250MG A134416-25G A134416-5G
77285-426EA 861.08 USD
77285-426 77285-428 77285-430 77285-432 77285-434 77285-436
4,5-Difluoro-2-iodobenzoic acid 98%
4,5-Difluoro-2-iodobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃F₂IO₂
MW: 284 g/mol
MDL Number: MFCD02684316
CAS Number: 130137-05-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR