5-(Trifluoromethyl)pyrimidin-2-amine 97%

Supplier: Ambeed
Warning

A134395-100MG A134395-250MG A134395-5G A134395-1G
77272-146EA 28.84 USD
77272-146 77272-148 77272-150 77566-634
5-(Trifluoromethyl)pyrimidin-2-amine 97%
5-(Trifluoromethyl)pyrimidin-2-amine

Precaución: For research use only.

Formula: C₅H₄F₃N₃
MW: 163.101 g/mol
MDL Number: MFCD10696951
CAS Number: 69034-08-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR