2-(4-Methylpiperidin-1-yl)-5-nitroaniline 95%

Supplier: Ambeed
Warning

A134303-100MG A134303-1G A134303-250MG
77234-646EA 29 USD
77234-646 77234-648 77234-650
2-(4-Methylpiperidin-1-yl)-5-nitroaniline 95%
2-(4-Methylpiperidin-1-yl)-5-nitroaniline

Precaución: for Research Use Only

Formula: C₁₂H₁₇N₃O₂
MDL Number: MFCD17020518
CAS Number: 188604-99-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR