6-Chloro-2-(trichloromethyl)-4H-chromen-4-one 97%

Supplier: Ambeed
Warning

A134156-10MG A134156-1MG A134156-25MG A134156-50MG A134156-5MG
77272-128EA 109.18 USD
77272-128 77272-130 77272-132 77272-134 77272-136
6-Chloro-2-(trichloromethyl)-4H-chromen-4-one 97%
6-Chloro-2-(trichloromethyl)-4H-chromen-4-one

Precaución: for Research Use Only

Formula: C₁₀H₄Cl₄O₂
MW: 297.94 g/mol
MDL Number: MFCD00760793
CAS Number: 133406-29-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR