ONO-4059 analog ≥98%

Supplier: Ambeed
Warning

Synonyms: (S)-9-(1-Acryloylpiperidin-3-yl)-6-amino-7-(4-phenoxyphenyl)-7,9-dihydro-8H-purin-8-one, 6-​Amino-​7,​9-​dihydro-​9-​[(3S)​-​1-​(1-​oxo-​2-​propen-​1-​yl)​-​3-​piperidinyl]​-​7-​(4-​phenoxyphenyl)​-8H-​purin-​8-​one

A133954-10MG A133954-1MG A133954-5MG
77234-634EA 105.06 USD
77234-634 77234-636 77234-638
ONO-4059 analog ≥98%
ONO-4059 analog

Precaución: for Research Use Only

Formula: C₂₅H₂₄N₆O₃
MW: 456.51 g/mol
MDL Number: MFCD29049511
CAS Number: 1351635-67-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR