4-Aminoindole 97%

Supplier: Ambeed
Warning

Synonyms: Indol-4-ylamine, 4-Indolamine, 4-Amino-1H-indole

A133822-1G A133822-250MG A133822-25G A133822-5G
77246-650EA 15 USD
77246-650 77246-652 77246-654 77246-656
4-Aminoindole 97%
4-Aminoindole

Precaución: for Research Use Only

Formula: C₈H₈N₂
MW: 132.16 g/mol
Melting Pt: 108
MDL Number: MFCD01076559
CAS Number: 5192-23-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR