4-(2H-1,2,3-Triazol-2-yl)benzaldehyde 97%

Supplier: Ambeed
Warning

A133650-100MG A133650-1G A133650-250MG
77285-404EA 83 USD
77285-404 77285-406 77285-408
4-(2H-1,2,3-Triazol-2-yl)benzaldehyde 97%
4-(2H-1,2,3-Triazol-2-yl)benzaldehyde

Precaución: for Research Use Only

Formula: C₉H₇N₃O
MDL Number: MFCD08669904
CAS Number: 179056-04-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR