3-Bromo-5-(chloromethyl)-2-fluoropyridine 95%

Supplier: Ambeed
Warning

A133605-100MG A133605-1G A133605-250MG
77234-624EA 42 USD
77234-624 77234-626 77234-628
3-Bromo-5-(chloromethyl)-2-fluoropyridine 95%
3-Bromo-5-(chloromethyl)-2-fluoropyridine

Precaución: for Research Use Only

Formula: C₆H₄BrClFN
MDL Number: MFCD16608114
CAS Number: 1227584-17-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR