2-Bromospiro[9H-fluorene-9,9'-[9H]xanthene] 97%

Supplier: Ambeed
Warning

Synonyms: 2-Bromospiro[fluorene-9,9'-xanthene]

A133582-10G A133582-1G A133582-250MG A133582-25G A133582-5G
77234-612EA 49 USD
77234-612 77234-614 77234-616 77234-618 77234-620
2-Bromospiro[9H-fluorene-9,9'-[9H]xanthene] 97%
2-Bromospiro[9H-fluorene-9,9'-[9H]xanthene]

Precaución: for Research Use Only

Formula: C₂₅H₁₅BrO
MDL Number: MFCD29050113
CAS Number: 899422-06-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR