4-(Pyridin-4-yl)thiazole-2-thiol 97%

Supplier: Ambeed
Warning

A133514-1G A133514-25G A133514-5G
77285-386EA 17.51 USD
77285-386 77285-388 77285-390 77285-392
4-(Pyridin-4-yl)thiazole-2-thiol 97%
4-(Pyridin-4-yl)thiazole-2-thiol

Precaución: for Research Use Only

Formula: C₈H₆N₂S₂
MDL Number: MFCD11506654
CAS Number: 77168-63-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR