3-Methyl-1,3-butanediol 95%

Supplier: Ambeed
Warning

A133476-100G A133476-500G A133476-5G A133476-25G
77272-096EA 35 USD
77272-096 77272-098 77272-100 77549-600
3-Methyl-1,3-butanediol 95%
3-Methyl-1,3-butanediol

Precaución: for Research Use Only

Formula: C₅H₁₂O₂
MDL Number: MFCD00059655
CAS Number: 2568-33-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR