Benzyl (2-(4-((4-fluorobenzyl)carbamoyl)-5-hydroxy-6-oxo-1,6-dihydropyrimidin-2-yl)propan-2-yl)carbamate 97%

Supplier: Ambeed
Warning

A133159-1G A133159-250MG
77246-624EA 18.54 USD
77246-624 77246-626 77246-628
Benzyl (2-(4-((4-fluorobenzyl)carbamoyl)-5-hydroxy-6-oxo-1,6-dihydropyrimidin-2-yl)propan-2-yl)carbamate 97%
Benzyl (2-(4-((4-fluorobenzyl)carbamoyl)-5-hydroxy-6-oxo-1,6-dihydropyrimidin-2-yl)propan-2-yl)carbamate

Precaución: for Research Use Only

Formula: C₂₃H₂₃FN₄O₅
MW: 454.45 g/mol
MDL Number: MFCD30489073
CAS Number: 519028-33-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR