3-Chloro-5-methylphenol 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Chloro-m-cresol, 3-Chloro-5-hydroxytoluene
A133129-10G
A133129-5G
77272-094EA
41
USD
77272-094
77747-950
3-Chloro-5-methylphenol 98%
3-Chloro-5-methylphenol
Precaución: for Research Use Only
Formula:
C₇H₇ClO |
MDL Number:
MFCD08234650 CAS Number: 58291-77-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...