3,5-Difluoro-4-iodoaniline 95%

Supplier: Ambeed
Warning

A132960-1G A132960-25G A132960-5G
77272-084EA 10.3 USD
77272-084 77272-086 77272-088
3,5-Difluoro-4-iodoaniline 95%
3,5-Difluoro-4-iodoaniline

Precaución: for Research Use Only

Formula: C₆H₄F₂IN
MW: 255.004 g/mol
MDL Number: MFCD07774190
CAS Number: 1542-34-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR