2,6-Dimethoxypyrazine 98%

Supplier: Ambeed
Warning

A132842-100G A132842-1G A132842-25G A132842-5G
77246-612EA 132 USD
77246-612 77246-614 77246-616 77246-618
2,6-Dimethoxypyrazine 98%
2,6-Dimethoxypyrazine

Precaución: for Research Use Only

Formula: C₆H₈N₂O₂
MDL Number: MFCD00955843
CAS Number: 4774-15-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR