1,3,3-Trimethyl-6'-(piperidin-1-yl)spiro[indoline-2,3'-naphtho[2,1-b][1,4]oxazine] 97%

Supplier: Ambeed
Warning

Synonyms: Technocolor Purple 3

A132795-100G A132795-1G A132795-25G A132795-5G A132795-250MG
77246-608EA 1161.84 USD
77246-608 77246-610 77615-058 77615-060 77745-950
1,3,3-Trimethyl-6'-(piperidin-1-yl)spiro[indoline-2,3'-naphtho[2,1-b][1,4]oxazine] 97%
1,3,3-Trimethyl-6'-(piperidin-1-yl)spiro[indoline-2,3'-naphtho[2,1-b][1,4]oxazine]

Precaución: for Research Use Only

Formula: C₂₇H₂₉N₃O
MW: 411.53 g/mol
MDL Number: MFCD00329486
CAS Number: 114747-45-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR