2-Ethoxyethyl methacrylate 98% stabilised

Supplier: Ambeed
Warning

Synonyms: Methacrylic acid 2-Ethoxyethyl ester, Ethylene glycol monoethyl ether Methacrylate

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

A132696-100G A132696-25G A132696-500G
77258-534EA 27 USD
77258-534 77258-536 77258-538
2-Ethoxyethyl methacrylate 98% stabilised
2-Ethoxyethyl methacrylate

Precaución: for Research Use Only

Formula: C₈H₁₄O₃
MW: 158.1950 g/mol
MDL Number: MFCD00009249
CAS Number: 2370-63-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR