4'-Formylbenzo-15-crown-5 (4'-formylbenzo-1,4,7,10,13-pentaoxacyclopentadecane) 98%

Supplier: Ambeed
Warning

Synonyms: 4'-Formylbenzo-15-crown 5-Ether, 2,3-(4-Formylbenzo)-1,4,7,10,13-pentaoxacyclopentadec-2-ene, 2,5,8,11,14-Pentaoxabicyclo[13.4.0]nonadeca-1(15),16,18-triene-17-carbaldehyde

A132688-100MG A132688-1G A132688-250MG
77246-602EA 37 USD
77246-602 77246-604 77246-606
4'-Formylbenzo-15-crown-5 (4'-formylbenzo-1,4,7,10,13-pentaoxacyclopentadecane) 98%
4'-Formylbenzo-15-crown-5 (4'-Formylbenzo-1,4,7,10,13-pentaoxacyclopentadecane)

Precaución: for Research Use Only

Formula: C₁₅H₂₀O₆
MW: 296.32 g/mol
MDL Number: MFCD00192180
CAS Number: 60835-73-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR