2,2,2-Trifluoroethyl butyrate 98%

Supplier: Ambeed
Danger

Synonyms: Butyric acid 2,2,2-Trifluoroethyl ester

A132457-1G A132457-25G A132457-5G
77246-588EA 9 USD
77246-588 77246-590 77246-592
2,2,2-Trifluoroethyl butyrate 98%
2,2,2-Trifluoroethyl butyrate

Precaución: for Research Use Only

Formula: C₆H₉F₃O₂
MDL Number: MFCD00077609
CAS Number: 371-27-7
UN: 3272
ADR: 3,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR