trans-3,4-Difluorocinnamic acid 97%

Supplier: Ambeed
Warning

A132412-100G A132412-1G A132412-25G A132412-5G
77258-528EA 94 USD
77258-528 77258-530 77258-532 77747-945
trans-3,4-Difluorocinnamic acid 97%
trans-3,4-Difluorocinnamic acid

Precaución: for Research Use Only

Formula: C₉H₆F₂O₂
MDL Number: MFCD00010320
CAS Number: 112897-97-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR