4-Nitro-L-phenylalanine monohydrate 97%

Supplier: Ambeed
Warning

Synonyms: (S)-2-Amino-3-(4-nitrophenyl)propanoic acid monohydrate, (S)-4-Nitrophenylalanine monohydrate, H-Phe(4-NO2)-OH.H2O

A132318-100G A132318-10G A132318-25G A132318-500G A132318-5G
77246-578EA 49.44 USD
77246-578 77246-580 77246-582 77246-584 77246-586
4-Nitro-L-phenylalanine monohydrate 97%
4-Nitro-L-phenylalanine monohydrate

Precaución: for Research Use Only

Formula: C₉H₁₂N₂O₅
MW: 228.20 g/mol
MDL Number: MFCD00150543
CAS Number: 207591-86-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR