6-Chloro-1H-indol-4-amine 98%

Supplier: Ambeed
Warning

A132285-100MG A132285-1G A132285-250MG
77285-360EA 27.81 USD
77285-360 77285-362 77285-364
6-Chloro-1H-indol-4-amine 98%
6-Chloro-1H-indol-4-amine

Precaución: for Research Use Only

Formula: C₈H₇ClN₂
MW: 166.61 g/mol
MDL Number: MFCD07781455
CAS Number: 431046-15-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR