1-Methyl-1H-pyrazolo[3,4-b]pyridin-3-ylamine 97%

Supplier: Ambeed
Danger

A132176-250MG A132176-1G
77234-576EA 15.45 USD
77234-576 77745-949
1-Methyl-1H-pyrazolo[3,4-b]pyridin-3-ylamine 97%
1-Methyl-1H-pyrazolo[3,4-b]pyridin-3-ylamine

Precaución: for Research Use Only

Formula: C₇H₈N₄
MW: 148.17 g/mol
MDL Number: MFCD04117796
CAS Number: 72583-83-6
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR