2-Amino-6-chloro-N-(2,4-dimethylphenyl)benzenesulfonamide 97%

Supplier: Ambeed
Warning

A132109-100G A132109-10G A132109-25G A132109-5G
77258-518EA 81.37 USD
77258-518 77258-520 77258-522 77258-524
2-Amino-6-chloro-N-(2,4-dimethylphenyl)benzenesulfonamide 97%
2-Amino-6-chloro-N-(2,4-dimethylphenyl)benzenesulfonamide

Precaución: for Research Use Only

Formula: C₁₄H₁₅ClN₂O₂S
MW: 310.80 g/mol
MDL Number: MFCD30749336
CAS Number: 1308061-19-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR