(R)-4-(1-Aminoethyl)phenol hydrochloride 97%

Supplier: Ambeed
Warning

A132032-100MG A132032-250MG
77246-574EA 130.81 USD
77246-574 77246-576
(R)-4-(1-Aminoethyl)phenol hydrochloride 97%
(R)-4-(1-Aminoethyl)phenol hydrochloride

Precaución: for Research Use Only

Formula: C₂₀H₁₄O₄
MW: 173.64 g/mol
MDL Number: MFCD08457901
CAS Number: 2061996-43-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR