3-(4-Methoxyphenyl)-5-isoxazolecarboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: 3-(4-Methoxyphenyl)isoxazole-5-carboxylic acid

A131955-1G A131955-5G
77272-048EA 49.44 USD
77272-048 77272-050
3-(4-Methoxyphenyl)-5-isoxazolecarboxylic acid 95%
3-(4-Methoxyphenyl)-5-isoxazolecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₉NO₄
MW: 219.1900 g/mol
MDL Number: MFCD04124288
CAS Number: 618383-47-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR