5-Methylbenzimidazole 98%

Supplier: Ambeed
Warning

A131930-100G A131930-10G A131930-1G A131930-25G A131930-5G
77246-562EA 120 USD
77246-562 77246-564 77246-566 77246-568 77246-570
5-Methylbenzimidazole 98%
5-Methylbenzimidazole

Precaución: for Research Use Only

Formula: C₈H₈N₂
MDL Number: MFCD00010740
CAS Number: 614-97-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR