4-Methoxy-3-(trifluoromethyl)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (4-Methoxy-3-(trifluoromethyl)phenyl)boronic acid, 4-Methoxy-3-trifluoromethylphenylboronic acid, 4-Methoxy-3-(trifluoromethyl)benzeneboronic acid

A131782-10G A131782-1G A131782-250MG A131782-5G
77234-568EA 192 USD
77234-568 77234-570 77234-572 77234-574
4-Methoxy-3-(trifluoromethyl)phenylboronic acid 98%
4-Methoxy-3-(trifluoromethyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₈BF₃O₃
MW: 219.95 g/mol
MDL Number: MFCD07363790
CAS Number: 149507-36-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR