4-(1,2,2-Triphenylvinyl)phenol 97%

Supplier: Ambeed
Warning

Synonyms: 1-(4-Hydroxyphenyl)-1,2,2-triphenylethene, 4-(1,2,2-Triphenylethenyl)phenol

A131706-100MG A131706-1G A131706-250MG A131706-25G A131706-5G
77234-552EA 6.18 USD
77234-552 77234-554 77234-556 77234-558 77234-560
4-(1,2,2-Triphenylvinyl)phenol 97%
4-(1,2,2-Triphenylvinyl)phenol

Precaución: for Research Use Only

Formula: C₂₆H₂₀O
MW: 348.45 g/mol
MDL Number: MFCD30342417
CAS Number: 76115-06-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR