4,5-Dimethylimidazole hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: 4,5-Dimethyl-1H-imidazole hydrochloride

A131658-250MG A131658-5G
77285-348EA 17.51 USD
77285-348 77285-350
4,5-Dimethylimidazole hydrochloride 95%
4,5-Dimethylimidazole hydrochloride

Precaución: for Research Use Only

Formula: C₅H₈N₂·ClH
MW: 132.591 g/mol
MDL Number: MFCD09743540
CAS Number: 53316-51-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR