3-(Difluoromethoxy)benzonitrile 97%

Supplier: Ambeed
Warning

A131641-100MG A131641-1G A131641-250MG A131641-25G A131641-5G
77234-542EA 5 USD
77234-542 77234-544 77234-546 77234-548 77234-550
3-(Difluoromethoxy)benzonitrile 97%
3-(Difluoromethoxy)benzonitrile

Precaución: for Research Use Only

Formula: C₈H₅F₂NO
MDL Number: MFCD00236225
CAS Number: 97582-88-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR