8-Chloroquinoline 98%

Supplier: Ambeed
Warning

A131578-100G A131578-10G A131578-1G A131578-25G A131578-5G
77258-482EA 188.49 USD
77258-482 77258-484 77258-486 77258-488 77258-490
8-Chloroquinoline 98%
8-Chloroquinoline

Precaución: for Research Use Only

Formula: C₉H₆ClN
MDL Number: MFCD00047618
CAS Number: 611-33-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR