3-[4-(Methylsulfanyl)phenyl]acrylic acid 98%

Supplier: Ambeed
Warning

A131564-1G A131564-250MG A131564-5G
77272-030EA 26 USD
77272-030 77272-032 77272-034
3-[4-(Methylsulfanyl)phenyl]acrylic acid 98%
3-[4-(Methylsulfanyl)phenyl]acrylic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₂S
MDL Number: MFCD00266583
CAS Number: 102016-58-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR