L-Prolinamide hydrochloride 96%

Supplier: Ambeed
Warning

Synonyms: (S)-Pyrrolidine-2-carboxamide hydrochloride, H-Pro-NH₂·HCl

A131495-100G A131495-10G A131495-1G A131495-5G A131495-25G
77285-342EA 114.33 USD
77285-342 77285-344 77285-346 77615-038 77747-937
L-Prolinamide hydrochloride 96%
L-Prolinamide hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₁ClN₂O
MW: 150.606 g/mol
MDL Number: MFCD00058156
CAS Number: 42429-27-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR