2-Methyl-6-(trifluoromethyl)pyrimidin-4-ol 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Hydroxy-2-methyl-6-(trifluoromethyl)pyrimidine, 2-Methyl-6-(trifluoromethyl)-4-pyrimidinol
A131347-1G
A131347-25G
A131347-5G
77285-330EA
5.15
USD
77285-330
77285-332
77285-334
2-Methyl-6-(trifluoromethyl)pyrimidin-4-ol 97%
2-Methyl-6-(trifluoromethyl)pyrimidin-4-ol
Precaución: for Research Use Only
Formula:
C₆H₅F₃N₂O MW: 178.1119 g/mol |
MDL Number:
MFCD03265321 CAS Number: 2836-44-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...