N-(3-Hydroxypyridin-2-yl)acetamide 97%

Supplier: Ambeed
Warning

Synonyms: 2-Acetamido-3-hydroxypyridine

A131327-1G A131327-250MG
77234-536EA 25.75 USD
77234-536 77234-538
N-(3-Hydroxypyridin-2-yl)acetamide 97%
N-(3-Hydroxypyridin-2-yl)acetamide

Precaución: for Research Use Only

Formula: C₇H₈N₂O₂
MW: 152.15 g/mol
MDL Number: MFCD09757484
CAS Number: 31354-48-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR